Idex MicroTight® Ferrule Plug, Black PEEK, 5/16-24 Coned; 1/EA
Representative image only.
Idex MicroTight® Ferrule Plug, Black PEEK, 5/16-24 Coned; 1/EA
Idex | Mfr: P-116| Item: 0201919
Make superior fingertight connections with capillary tubing
₹5,600.00INR / Each
₹5,600.00INR / Each
Product Description
Key Features
  • Constructed of durable PEEK
  • Use to close a port
  • 1 year warranty
More About this Item
Select microferrules for 360 µm, 1/32", 1/16" OD tubing or MicroTight tubing sleeves.
1 Year Warranty
Specifications
  • MaterialPEEK
  • ColorBlack
  • Compatible With5/16-24 coned
  • DescriptionMicroTight® Ferrule Plug, Black PEEK, 5/16-24 Coned; 1/EA