Idex® LuerTight™ Fittings
Convenience of quick connect and disconnect of a luer connection while holding tubing firmly in place
  • Manufactured from inert polymers
LuerTight™ fittings are designed to connect fluoropolymer tubing without the use of barbs or nuts by integrating ferrules into the polypropylene bodies, eliminating the extra bulk and expense of added fittings.
480.00 - ₹5,600.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Connection Type
Availability Pricing
Idex LuerTight™ Fitting, Female, Natural ETFE, 1/8" OD Tubing; 1/EA 0202389 Mfr # P-830
COMPARE
1/8 ETFE HPLC/Chromatography
₹1,500.00INR / Each
Idex LuerTight™ Ferrule, Natural ETFE, 1/8" OD Tubing; 1/EA 0202390 Mfr # P-830-02
COMPARE
1/8 ETFE HPLC/Chromatography
₹480.00INR / Each
Idex LuerTight™ Set Plug, Natural ETFE, 1/16" and 1/8" OD Tubing; 1/EA 0202391 Mfr # P-830T
COMPARE
1/8 ETFE HPLC/Chromatography
₹4,200.00INR / Each
Idex LuerTight™ Fitting, Male, Natural ETFE, 1/8" OD Tubing; 1/EA 0202392 Mfr # P-831
COMPARE
1/8 ETFE HPLC/Chromatography
₹1,500.00INR / Each
Idex LuerTight™ Fitting, Female, Natural ETFE, 1/16" OD Tubing; 1/EA 0202393 Mfr # P-835
COMPARE
1/16 ETFE HPLC/Chromatography
₹1,500.00INR / Each
Idex LuerTight™ Fitting, Male, Natural ETFE, 1/16" OD Tubing; 1/EA 0202394 Mfr # P-836
COMPARE
1/16 ETFE HPLC/Chromatography
₹1,500.00INR / Each
Idex LuerTight™ Ferrule, Yellow ETFE, 1/16" OD Tubing; 1/EA 0202395 Mfr # P-836-02
COMPARE
1/16 ETFE HPLC/Chromatography
₹800.00INR / Each
Idex LuerTight™ Fitting System, Natural Polypropylene/ETFE, 1/16" OD Tubing; 1/EA 0202396 Mfr # P-837
COMPARE
1/16 ETFE/Polypropylene HPLC/Chromatography
₹5,600.00INR / Each
Idex LuerTight™ Fitting System, Natural Polypropylene/ETFE, 1/8" OD Tubing; 1/EA 0202397 Mfr # P-838
COMPARE
1/8 ETFE/Polypropylene HPLC/Chromatography
₹5,600.00INR / Each
Showing 1- 9 of 9