Idex LuerTight™ Ferrule, Yellow ETFE, 1/16" OD Tubing; 1/EA
Representative image only.
Idex LuerTight™ Ferrule, Yellow ETFE, 1/16" OD Tubing; 1/EA
Idex | Mfr: P-836-02| Item: 0202395
Convenience of quick connect and disconnect of a luer connection while holding tubing firmly in place
₹800.00INR / Each
₹800.00INR / Each
Product Description
Key Features
  • Manufactured from inert polymers
  • Yellow ETFE ferrule for 1/16" OD tubing LuerTight fitting systems 02023-96
  • 1 year warranty
More About this Item
LuerTight™ fittings are designed to connect fluoropolymer tubing without the use of barbs or nuts by integrating ferrules into the polypropylene bodies, eliminating the extra bulk and expense of added fittings.
1 Year Warranty
Specifications
  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Compression Size / Tube Od (Port 1) (mm)1.6
  • ColorYellow
  • DescriptionLuerTight™ Ferrule, Yellow ETFE, 1/16" OD Tubing; 1/EA