Idex LuerTight™ Ferrule, Natural ETFE, 1/8" OD Tubing; 1/EA
Representative image only.
Idex LuerTight™ Ferrule, Natural ETFE, 1/8" OD Tubing; 1/EA
Idex | Mfr: P-830-02| Item: 0202390
Convenience of quick connect and disconnect of a luer connection while holding tubing firmly in place
₹480.00INR / Each
₹480.00INR / Each
Product Description
Key Features
  • Manufactured from inert polymers
  • Natural ETFE ferrule for 1/8" OD tubing LuerTight fitting systems 02023-97
  • 1 year warranty
More About this Item
LuerTight™ fittings are designed to connect fluoropolymer tubing without the use of barbs or nuts by integrating ferrules into the polypropylene bodies, eliminating the extra bulk and expense of added fittings.
1 Year Warranty
Specifications
  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Compression Size / Tube Od (Port 1) (in)1/8
  • Compression Size / Tube Od (Port 1) (mm)3.2
  • ColorNatural
  • DescriptionLuerTight™ Ferrule, Natural ETFE, 1/8" OD Tubing; 1/EA