Idex LuerTight™ Set Plug, Natural ETFE, 1/16" and 1/8" OD Tubing; 1/EA
Representative image only.
Idex LuerTight™ Set Plug, Natural ETFE, 1/16" and 1/8" OD Tubing; 1/EA
Idex | Mfr: P-830T| Item: 0202391
Convenience of quick connect and disconnect of a luer connection while holding tubing firmly in place
₹4,200.00INR / Each
₹4,200.00INR / Each
Product Description
Key Features
  • Manufactured from inert polymers
  • Used to swage ferrules into LuerTight fittings
  • For female nuts 02023-93 and 02023-91
  • 1 year warranty
More About this Item
LuerTight™ fittings are designed to connect fluoropolymer tubing without the use of barbs or nuts by integrating ferrules into the polypropylene bodies, eliminating the extra bulk and expense of added fittings.
1 Year Warranty
Specifications
  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Compression Size / Tube Od (Port 1) (in)1/8
  • Compression Size / Tube Od (Port 1) (mm)3.2
  • ColorNatural
  • DescriptionLuerTight™ Set Plug, Natural ETFE, 1/16" and 1/8" OD Tubing; 1/EA