IDEX® Inline Solvent MicroFilters
Protect your columns from particulate contamination
  • Withstands up to 5000 psi (345 bar)
Specifically engineered inline solvent MicroFilters prevent particulate contamination ensuring reliable chromatographic results even at nanoliter flow rates. Filters protect columns from particles often originating in the mobile phase or sample, or from pump seal and sample injection valve wear. Connect directly using 1/32" OD tubing or choose MicroTight® tubing sleeve models for use with capillary tubing.
38,600.00 - ₹112,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connections Membrane Material Pore Size (µm)
Availability Pricing
Idex M-135-1 Inline Microfilter Assembly, SS, 1 µm SS Screen, MicroTight® Tubing Sleeves; 1/EA 4271129 Mfr # M-135-1
COMPARE
MicroTight® tubing sleeves Stainless Steel 1
₹38,600.00INR / Each
Idex M-520 Inline Microfilter Assembly, SS, 0.5 µm PEEK Frit, MicroTight® Tubing Sleeves; 1/EA 4271131 Mfr # M-520
COMPARE
MicroTight® tubing sleeves PEEK 0.5
₹53,600.00INR / Each
Idex M-525 Inline Microfilter Assembly, PCTFE, 0.5 µm, PEEK Frit, 1/32" OD Tubing; 1/EA 4271133 Mfr # M-525
COMPARE
1/32" OD tubing PEEK 0.5
₹46,900.00INR / Each
Idex M-547 Inline Mini Microfilter Assembly, PEEK, 1 µm SS Frit, 1/32" OD Tubing; 1/EA 4271137 Mfr # M-547
COMPARE
1/32" OD tubing Stainless Steel 1
₹112,000.00INR / Each