Heavy-Duty Forceps, PTFE
Totally inert and non-contaminating PTFE Teweezer Forceps
Chemical-resistant, noncontaminating PTFE forceps are ideal for use in all applications. Heat resistant up to 572°F (300°C) for short periods and up to 500°F (260°C) consistently.
11,400.00 - ₹15,500.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Overall Length (in)
Availability Pricing
Dynalon Square-Tip Autoclavable Heavy-Duty PTFE Forceps, 4" L; 1/Pk 0644202 Mfr # 316504-0000
COMPARE
PTFE 4
₹11,400.00INR / Each
Dynalon Heavy Duty PTFE Forceps Square End 7 7/8" 0644206 Mfr # 316504-0002
COMPARE
PTFE 7 7/8
₹15,500.00INR / Each
Dynalon Pointed-Tip Autoclavable Heavy-Duty PTFE Forceps, 5-7/8" L; 1/Pk 0644210 Mfr # 316514-0001
COMPARE
PTFE 5 7/8
₹13,800.00INR / Each