Dynalon Heavy Duty PTFE Forceps Square End 7 7/8"
Representative image only.
Dynalon Heavy Duty PTFE Forceps Square End 7 7/8"
Dynalon | Mfr: 316504-0002| Item: 0644206
Totally inert and non-contaminating PTFE Teweezer Forceps
₹15,500.00INR / Each
₹15,500.00INR / Each
Product Description
More About this Item
Chemical-resistant, noncontaminating PTFE forceps are ideal for use in all applications. Heat resistant up to 572°F (300°C) for short periods and up to 500°F (260°C) consistently.
Specifications
  • Overall Length (in)7 7/8
  • Overall Length (cm)20
  • MaterialPTFE
  • SterileNo
  • DescriptionHeavy Duty PTFE Forceps Square End 7 7/8"