Dynalon Pointed-Tip Autoclavable Heavy-Duty PTFE Forceps, 5-7/8" L; 1/Pk
Representative image only.
Dynalon Pointed-Tip Autoclavable Heavy-Duty PTFE Forceps, 5-7/8" L; 1/Pk
Dynalon | Mfr: 316514-0001| Item: 0644210
Totally inert and non-contaminating PTFE Teweezer Forceps
₹13,800.00INR / Each
₹13,800.00INR / Each
Product Description
More About this Item
Chemical-resistant, noncontaminating PTFE forceps are ideal for use in all applications. Heat resistant up to 572°F (300°C) for short periods and up to 500°F (260°C) consistently.
Specifications
  • Overall Length (in)5 7/8
  • Overall Length (cm)14.92
  • MaterialPTFE
  • SterileNo
  • DescriptionPointed-Tip Autoclavable Heavy-Duty PTFE Forceps, 5-7/8" L; 1/Pk