H-B Instrument Enviro-Safe Easy-Read Nonhazardous Glass Thermometers Traceable to NIST - Celsius Scale
Each thermometer is individually calibrated
  • Celsius-scale
  • Designed to be very easy to read
  • Replace mercury thermometers with the same accuracy without any hazards
  • 100% nontoxic, nonhazardous, biodegradable, and “green”
  • NIST-Certified detailed, laminated report of calibration
Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read. These thermometers are also individually calibrated to NIST standards at specific temperature and supplied with an NIST report of calibration stating corrections to one-tenth the smallest division. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. These thermometers come complete with ring top anti-roll fitting, individual serial number, NIST report of calibration, recyclable packaging, and are RoHS compliant.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Overall Length (mm)
Availability Pricing
H-B Instrument B60202-0100 Easy-Read Individually Calibrated Liquid-In-Glass Thermometer; -20 to 110C, 76mm Immersion, Environmentally Friendly 0800848 Mfr # B602020100
COMPARE
110 -20 300
Call For Price
H-B Instrument B60202-0900 Easy-Read Individually Calibrated Liquid-In-Glass Thermometer; -10 to 225C, Total Immersion, Environmentally Friendly 0800854 Mfr # B602020900
COMPARE
225 -10 350
Call For Price
H-B Instrument B60202-1300 Easy-Read Individually Calibrated Liquid-In-Glass Thermometer; -10 to 260C, 76mm Immersion, Environmentally Friendly 9030078 Mfr # B602021300
COMPARE
260 -10 350
Call For Price
H-B Instrument B60202-1200 Easy-Read Individually Calibrated Liquid-In-Glass Thermometer; 0 to 300F, 76 mm Immersion, Environmentally Friendly 9031003 Mfr # B602021200
COMPARE
300
Call For Price
H-B Instrument B60204-0000 Enviro-Safe Individually Calibrated Precision Liquid-In-Glass Thermometer; -1 to 61C, 76mm Immersion, Environmentally Friendly 9031006 Mfr # B602040000
COMPARE
61 -1 420
Call For Price
H-B Instrument B60206-0000 Enviro-Safe Calibrated Dry Block/Incubator Liquid-In-Glass Thermometer; 24 to 57C, 35mm Immersion, Enviro-Safe Non-Toxic Liquid Fill 9031007 Mfr # B602060000
COMPARE
57 24 175