H-B Instrument B60204-0000 Enviro-Safe Individually Calibrated Precision Liquid-In-Glass Thermometer; -1 to 61C, 76mm Immersion, Environmentally Friendly

Representative image only.

H-B Instrument B60204-0000 Enviro-Safe Individually Calibrated Precision Liquid-In-Glass Thermometer; -1 to 61C, 76mm Immersion, Environmentally Friendly

H-B Instrument | Mfr: B602040000| Item: 9031006

Each thermometer is individually calibrated

Call For Price

Call For Price
Product Description

Includes

Ring top anti-roll fitting, storage box, and laminated NIST- traceable calibration report

Key Features

  • Celsius-scale
  • Designed to be very easy to read
  • Replace mercury thermometers with the same accuracy without any hazards
  • 100% nontoxic, nonhazardous, biodegradable, and “green”
  • NIST-Certified detailed, laminated report of calibration
  • 1 year warranty

More About this Item

Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read. These thermometers are also individually calibrated to NIST standards at specific temperature and supplied with an NIST report of calibration stating corrections to one-tenth the smallest division. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. These thermometers come complete with ring top anti-roll fitting, individual serial number, NIST report of calibration, recyclable packaging, and are RoHS compliant.

1 Year Warranty
Specifications
  • Length (")16 1/2
  • Min Temperature (° C)-1
  • Max Temperature (° C)61
  • Accuracy±0.5°C
  • Division (° C)0.1
  • Thermometer FillGreen Spirit
  • Overall Length (InchShort)16.5
  • Overall Length (mm)420
  • Immersion Depth (mm)76
  • Stem MaterialWhite-Back Glass
  • Calibration Points0, 37, 56°C
  • DescriptionEnviro-Safe Individually Calibrated Precision Liquid-In-Glass Thermometer; -1 to 61C, 76mm Immersion, Environmentally Friendly
  • Warranty1 year
Reviews
Q&A