H-B Instrument B60202-0900 Easy-Read Individually Calibrated Liquid-In-Glass Thermometer; -10 to 225C, Total Immersion, Environmentally Friendly

Representative image only.

H-B Instrument B60202-0900 Easy-Read Individually Calibrated Liquid-In-Glass Thermometer; -10 to 225C, Total Immersion, Environmentally Friendly

H-B Instrument | Mfr: B602020900| Item: 0800854

Each thermometer is individually calibrated

Call For Price

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

Call For Price

Buy It Together

Current Item

H-B Instrument B60202-0900 Easy-Read Individually Calibrated Liquid-In-Glass Thermometer; -10 to 225C, Total Immersion, Environmentally Friendly
H-B Instrument B60202-0900 Easy-Read Individually Calibrated Liquid-In-Glass Thermometer; -10 to 225C, Total Immersion, Environmentally Friendly
0800854

Call For Price
+ =
Total Price 33100.00 ADD 2 ITEMS TO CART
Product Description

Includes

Ring top anti-roll fitting, storage box, and laminated NIST- traceable calibration report

Key Features

  • Celsius-scale
  • Designed to be very easy to read
  • Replace mercury thermometers with the same accuracy without any hazards
  • 100% nontoxic, nonhazardous, biodegradable, and “green”
  • NIST-Certified detailed, laminated report of calibration
  • 5 year warranty

More About this Item

Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read. These thermometers are also individually calibrated to NIST standards at specific temperature and supplied with an NIST report of calibration stating corrections to one-tenth the smallest division. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. These thermometers come complete with ring top anti-roll fitting, individual serial number, NIST report of calibration, recyclable packaging, and are RoHS compliant.

5 Year Warranty
Specifications
  • Length (")14
  • Min Temperature (° C)-10
  • Max Temperature (° C)225
  • Accuracy±1.0°C
  • Division (° C)1
  • Thermometer FillBlack Spirit
  • Overall Length (InchShort)14
  • Overall Length (mm)350
  • Immersion Depth (mm)350
  • Stem MaterialYellow-Back Glass
  • Calibration Points1, 100, 200°C
  • DescriptionEasy-Read Individually Calibrated Liquid-In-Glass Thermometer; -10 to 225C, Total Immersion, Environmentally Friendly
  • Warranty5 years
Reviews
Q&A