H-B Instrument B60206-0000 Enviro-Safe Calibrated Dry Block/Incubator Liquid-In-Glass Thermometer; 24 to 57C, 35mm Immersion, Enviro-Safe Non-Toxic Liquid Fill
Representative image only.
H-B Instrument B60206-0000 Enviro-Safe Calibrated Dry Block/Incubator Liquid-In-Glass Thermometer; 24 to 57C, 35mm Immersion, Enviro-Safe Non-Toxic Liquid Fill
H-B Instrument | Mfr: B602060000| Item: 9031007
Each thermometer is individually calibrated
Call For Price
Call For Price
Product Description
Includes
Ring top anti-roll fitting, storage box, and laminated NIST- traceable calibration report
Key Features
  • Celsius-scale
  • Designed to be very easy to read
  • Replace mercury thermometers with the same accuracy without any hazards
  • 100% nontoxic, nonhazardous, biodegradable, and “green”
  • NIST-Certified detailed, laminated report of calibration
  • 1 year warranty
More About this Item
Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read. These thermometers are also individually calibrated to NIST standards at specific temperature and supplied with an NIST report of calibration stating corrections to one-tenth the smallest division. All Enviro-Safe glass thermometers contain an environmentally friendly non-mercury liquid that is 100% certified as non-toxic, non-hazardous, and fully biodegradable. In case of breakage the liquid can be cleaned up easily with soap and water. These thermometers come complete with ring top anti-roll fitting, individual serial number, NIST report of calibration, recyclable packaging, and are RoHS compliant.
1 Year Warranty
Specifications
  • Length (")6 29/32
  • Min Temperature (° C)24
  • Max Temperature (° C)57
  • Accuracy0.5°C
  • Division (° C)0.5
  • Thermometer FillGreen Spirit
  • Overall Length (InchShort)6.9
  • Overall Length (mm)175
  • Immersion Depth (mm)35
  • Stem MaterialWhite-Back Glass
  • Calibration Points37°C
  • DescriptionEnviro-Safe Calibrated Dry Block/Incubator Liquid-In-Glass Thermometer; 24 to 57C, 35mm Immersion, Enviro-Safe Non-Toxic Liquid Fill