Glas-Col Tissue Homogenizing System Glass Tubes
Choose a tube that fits your pestle and sample size
  • Precision-bore borosilicate glass
  • Tubes can withstand temperatures as low as liquid nitrogen
Tubes are constructed of precision-bore borosilicate glass.
20,600.00 - ₹47,500.00INR / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Material Overall Length (mm)
Availability Pricing
Glas-Col 099C S30 Tissue Homogenizing System Glass Tube; 2 mL volume, straight side 0436814 Mfr # 099C S30
COMPARE
2 Glass 146
₹20,600.00INR / Each
Glas-Col 099C S31 Tissue Homogenizing System Glass Tube; 10 mL volume, straight side 0436817 Mfr # 099C S31
COMPARE
10 Glass 146
₹20,600.00INR / Each
Glas-Col 099C S35 Tissue Homogenizing System Glass Tube; 15 mL volume, straight side 0436818 Mfr # 099C S35
COMPARE
15 Glass 146
₹23,600.00INR / Each
Glas-Col 099C S38 Tissue Homogenizing System Glass Tube; 5 mL volume, straight side 0436819 Mfr # 099C S38
COMPARE
5 Glass 102
₹20,600.00INR / Each
Glas-Col 099C S32 Tissue Homogenizing System Glass Tube; 20 mL volume, straight side 0436821 Mfr # 099C S32
COMPARE
20 Glass 146
₹25,800.00INR / Each
Glas-Col 099C S37 Tissue Homogenizing System Glass Tube; 40 mL volume, straight side 04368-25 0436825 Mfr # 099C S37
COMPARE
40 Glass 146
₹33,900.00INR / Each
Glas-Col 099C S36 Tissue Homogenizing System Glass Tube; 15 mL volume, straight side 0436829 Mfr # 099C S36
COMPARE
15 Glass 146
₹31,400.00INR / Each
Glas-Col 099C S39 Tissue Homogenizing System Glass Tube; 40 mL volume, straight side 04368-31 0436831 Mfr # 099C S39
COMPARE
40 Glass 146
₹35,600.00INR / Each
Glas-Col 099C S71 Tissue Homogenizing System Glass Tube; 2 mL volume, tapered side 04368-33 0436833 Mfr # 099C S71
COMPARE
2 Glass 146
₹35,400.00INR / Each
Glas-Col 099C S74 Tissue Homogenizing System Glass Tube; 15 mL volume, tapered side 0436835 Mfr # 099C S74
COMPARE
15 Glass 146
₹39,600.00INR / Each
Glas-Col 099C S78 Tissue Homogenizing System Glass Tube; 40 mL volume, tapered side 0436839 Mfr # 099C S78
COMPARE
40 Glass 146
₹47,500.00INR / Each
Glas-Col 099C S73 Tissue Homogenizing System Glass Tube; 2 mL volume, tapered side 04368-41 0436841 Mfr # 099C S73
COMPARE
2 Glass 146
₹32,900.00INR / Each
Glas-Col 099C S75 Tissue Homogenizing System Glass Tube; 15 mL volume, tapered side 0436843 Mfr # 099C S75
COMPARE
15 Glass 146
₹42,100.00INR / Each
Showing 1- 13 of 13