Fischer Technology Ferrite Content Probes for FERITSCOPE® FMP30
Measure the delta ferrite content in austenitic and duplex steels
  • Plug-in magnetic induction probes
  • Measurement range: 0.1 to 80% Fe or 0.1 to 110 WRC-FN
Measuring according to the Basel standard Checking ferrite content in welding seams and austenitic cladding.
359,000.00 - ₹460,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Application
Availability Pricing
Fischer Technology FGAB1.3-Fe Straight Ferrite Content Probe for Flat and Curved Surfaces 5410016 Mfr # 604-264
COMPARE
Straight probe: for flat and curved surfaces
₹359,000.00INR / Each
Fischer Technology FGABW1.3-Fe Angled Ferrite Content Probe for Flat Surfaces, Pipes, and Gaps 5410017 Mfr # 604-337
COMPARE
Angled probe: for flat surfaces, pipes, and gaps
₹380,000.00INR / Each
Fischer Technology FGABI1.3-150-Fe Narrow Ferrite Content Probe for Measuring in Pipes and Grooves 5410018 Mfr # 604-254
COMPARE
Narrow probe: for measuring in pipes and grooves
₹460,000.00INR / Each