Fischer Technology FGABI1.3-150-Fe Narrow Ferrite Content Probe for Measuring in Pipes and Grooves
Representative image only.
Fischer Technology FGABI1.3-150-Fe Narrow Ferrite Content Probe for Measuring in Pipes and Grooves
Fischer Technology | Mfr: 604-254| Item: 5410018
Measure the delta ferrite content in austenitic and duplex steels
₹460,000.00INR / Each
₹460,000.00INR / Each
Product Description
Key Features
  • Plug-in magnetic induction probes
  • Measurement range: 0.1 to 80% Fe or 0.1 to 110 WRC-FN
  • Narrow probe fits easily into pipes and grooves
  • Use on 6 month warranty
More About this Item
Measuring according to the Basel standard Checking ferrite content in welding seams and austenitic cladding.
6 Month Warranty
Specifications
  • Compatible WithFERITSCOPE® FMP30
  • ApplicationNarrow probe: for measuring in pipes and grooves
  • DescriptionNarrow Ferrite Content Probe for Measuring in Pipes and Grooves