Fischer Technology FGABW1.3-Fe Angled Ferrite Content Probe for Flat Surfaces, Pipes, and Gaps
Representative image only.
Fischer Technology FGABW1.3-Fe Angled Ferrite Content Probe for Flat Surfaces, Pipes, and Gaps
Fischer Technology | Mfr: 604-337| Item: 5410017
Measure the delta ferrite content in austenitic and duplex steels
₹380,000.00INR / Each
₹380,000.00INR / Each
Product Description
Key Features
  • Plug-in magnetic induction probes
  • Measurement range: 0.1 to 80% Fe or 0.1 to 110 WRC-FN
  • Angled for easy use on flat surfaces, pipes, and gaps
  • 6 month warranty
More About this Item
Measuring according to the Basel standard Checking ferrite content in welding seams and austenitic cladding.
6 Month Warranty
Specifications
  • Compatible WithFERITSCOPE® FMP30
  • ApplicationAngled probe: for flat surfaces, pipes, and gaps
  • DescriptionAngled Ferrite Content Probe for Flat Surfaces, Pipes, and Gaps