Fischer Technology FGAB1.3-Fe Straight Ferrite Content Probe for Flat and Curved Surfaces
Representative image only.
Fischer Technology FGAB1.3-Fe Straight Ferrite Content Probe for Flat and Curved Surfaces
Fischer Technology | Mfr: 604-264| Item: 5410016
Measure the delta ferrite content in austenitic and duplex steels
₹359,000.00INR / Each
₹359,000.00INR / Each
Product Description
Key Features
  • Plug-in magnetic induction probes
  • Measurement range: 0.1 to 80% Fe or 0.1 to 110 WRC-FN
  • Use on flat and curved surfaces
  • 6 month warranty
More About this Item
Measuring according to the Basel standard Checking ferrite content in welding seams and austenitic cladding.
6 Month Warranty
Specifications
  • Compatible WithFERITSCOPE® FMP30
  • ApplicationStraight probe: for flat and curved surfaces
  • DescriptionStraight Ferrite Content Probe for Flat and Curved Surfaces