Extech Type-K Thermocouple Bead Wire Probes
Exposed tip allows for fast response time
  • Compatible with most multimeters and thermometers that have Type K temperature function
Use these bead wire probes to measure the temperature of air and gas.
5,300.00 - ₹12,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° C) Min Temperature (° C)
Availability Pricing
Extech Type K, Bead Wire Probe, Mini-Connector with Banana Plug Adapter 9041509 Mfr # TP873
COMPARE
Mini Connector with Banana Plug Adapter 300 -30
₹5,500.00INR / Each
Extech TP870 Type-K, Bead Wire Probe, Mini-Connector 9041510 Mfr # TP870
COMPARE
Mini Connector 250 -40
₹5,300.00INR / Each
Extech TP875 Type-K, Wide Temperature Range Bead Wire Probe, Mini-Connector with Banana Plug Adapter 9500128 Mfr # TP875
COMPARE
Mini Connector with Banana Plug Adapter 538 -40
₹12,000.00INR / Each