Extech Type K, Bead Wire Probe, Mini-Connector with Banana Plug Adapter
Representative image only.
Extech Type K, Bead Wire Probe, Mini-Connector with Banana Plug Adapter
Extech | Mfr: TP873| Item: 9041509
Exposed tip allows for fast response time
₹5,500.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹5,500.00INR / Each
Buy It Together
Current Item
Extech Type K, Bead Wire Probe, Mini-Connector with Banana Plug Adapter
Extech Type K, Bead Wire Probe, Mini-Connector with Banana Plug Adapter
9041509
₹5,500.00INR / Each
+ =
Total Price 38300.00 ADD 2 ITEMS TO CART
Product Description
Includes
Banana plug adapter.
Key Features
  • Compatible with most multimeters and thermometers that have Type K temperature function
  • Temperature range: -30 to 300°C (-22 to 572°F)
  • 0.9 m (36") long cable
  • 6 month warranty
More About this Item
Use these bead wire probes to measure the temperature of air and gas.
6 Month Warranty
Specifications
  • Probe TypeK
  • Min Temperature (° F)-20
  • Max Temperature (° F)572
  • Min Temperature (° C)-30
  • Max Temperature (° C)300
  • Response Time (Sec)2
  • Connector TypeMini Connector with Banana Plug Adapter
  • Cable TypeStraight
  • Cable Length (InchShort)36
  • DescriptionType K, Bead Wire Probe, Mini and Banana Connectors
  • Warranty6 months