Eppendorf epT.I.P.S Pipet Tips for Adjustable-Volume 2000 and 2100 Research Pipettes
Maintain accuracy even when dispensing liquids other than water
  • Original high-quality Eppendorf pipette tips
  • Packaged in resealable bags
  • Available in sizes from 10µL to 10 mL
Use these Eppendorf® pipette tips with the Fixed-Volume Reference series, or the Adjustable Volume 2000 or 2100 Research Pipettors. The epTIPS - Totally Integrated Pipetting System - have been perfectly matched to Eppendorf Pipettes. This results in consistently reduced tip attachment and ejection forces, while ensuring complete sealing. 200 µl, 300 µl and 1,000 µl tips are color coded, yellow and blue, respectively.
All Eppendorf pipettes in combination with Eppendorf tips satisfy the requirements of DIN 12 650-2 and of the imminent EN ISO 8655-2.
16,300.00 - ₹28,300.00INR / Case of 1000
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Volume (µL) Min Volume (µL) Package Of
Availability Pricing
Eppendorf 022492039 Pipette Tips, 200 uL, Bulk; 1000/Cs WW-07935-12 Mfr # 22492039
COMPARE
200 2 Bulk; 1000/cs
₹16,300.00INR / Case of 1000
Eppendorf 022492071 Pipette Tips, 2500 uL, Bulk; 5 Bags x 100 Tips WW-07935-22 Mfr # 22492071
COMPARE
2500 250 Bulk; 500/cs
₹20,700.00INR / Case of 500
Eppendorf 022491539 Pipette Tips, 200 uL; 10 Racks x 96 Tips WW-07935-32 Mfr # 022491539
COMPARE
200 2 10 Racks; 960/pk
₹19,100.00INR / Case of 960
Eppendorf 022492080 Pipette Tips, 100 to 5000 uL, Bulk; 5 Bags x 100 Tips WW-07935-63 Mfr # 022492080
COMPARE
5000 100 Bulk; 500/cs
₹28,300.00INR / Case of 500
Eppendorf 022492021 Pipette Tips, 0.5 to 20 uL, Bulk; 1000/Cs WW-07936-89 Mfr # 022492021
COMPARE
20 0.5 Bulk; 1000/cs
₹20,700.00INR / Case of 1000