Digi-Sense Thermocouple Magnetic / Dropping Surface Probes
Magnetic Probes Designed for Monitoring a Wide Range of Flat Surfaces
  • Easily attach to any ferrous surface
  • Ideal for long term installation
  • Magnet strength is 15lbs max
  • Response Time: 1.25 seconds or greater
  • Each probe comes with color coded ANSI coiled cables or connectors: type J-black, type K-yellow, type T-blue, and type E-purple
  • Custom designs or changes to this product are available upon request
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
28,700.00 - ₹29,900.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type Description
Availability Pricing
COMPARE
500 T Type-T Dropping / Magnetic Probe 1.5" L Mini Conn Exp 5ft Coil Cord
₹29,900.00INR / Each
COMPARE
1200 K Type-K Dropping / Magnetic Probe 1.5" L Mini Conn Exp 10ft SS Braid Cable
₹29,500.00INR / Each
COMPARE
1200 K Type-K Dropping / Magnetic Probe 1.5" L Mini Conn Exp 5ft Coil Cord
₹29,400.00INR / Each
COMPARE
1200 J Type-J Dropping / Magnetic Probe 1.5" L Mini Conn Exp 5ft Coil Cord
₹29,400.00INR / Each
COMPARE
1200 J Type-J Dropping / Magnetic Probe 1.5" L Mini Conn Exp 10ft SS Braid Cable
₹28,700.00INR / Each
COMPARE
500 T Type-T Dropping / Magnetic Probe 1.5" L Mini Conn Exp 10ft SS Braid Cable
₹28,700.00INR / Each