Digi-Sense Thermocouple Heavy-Duty Industrial Penetration Probes
Heavy-Duty Industrial Penetration Probes Are Designed For Soft and Semi-Soft Materials
  • Features ICONEL®600 sheath
  • Response Time: 2 seconds or greater
  • Probe comes with stainless steel handle.
  • Each probe comes with color coded ANSI miniconnectors: type J-black, type K-yellow, and type T-blue.
  • Custom designs or changes to this product are available upon request
Flex armor is not water tight and is not recommended for damp or aqueous environments.
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
17,300.00 - ₹17,700.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type Description
Availability Pricing
COMPARE
500 T Type-T Hvy-Dty Ind SS Handle Mini Conn 36" L .188 Dia Grd 3ft Armor Cable
₹17,300.00INR / Each
COMPARE
1700 K Type-K Hvy-Dty Ind SS Handle Mini Conn 36" L .188 Dia Grd 3ft Armor Cable
₹17,700.00INR / Each
COMPARE
1400 J Type-J Hvy-Dty Ind SS Handle Mini Conn 36" L .188 Dia Grd 3ft Armor Cable
₹17,600.00INR / Each