Digi-Sense Thermocouple Flexible Wire Probes - PVC-Insulated with Epoxy-Coated Tip
PVC Insulation with Epoxy-Coated Tip Used for Short Term Immersion
  • Epoxy-coated tip can be used for applications requiring an isolated junction.
  • Response Time: 1 seconds or greater
  • All insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue.
  • Custom designs or changes to this product are available upon request
Exposed-junction probes are not recommended for use with aqueous solutions.
Do not subject type J exposed-junction probes to moisture as they will rust
5,900.00 - ₹6,900.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type Sheath Length (in)
Availability Pricing
COMPARE
221 J 120
₹6,100.00INR / Each
COMPARE
221 K 120
₹5,900.00INR / Each
COMPARE
221 T 120
₹5,900.00INR / Each
COMPARE
221 T 60
₹6,900.00INR / Each