Digi-Sense Thermocouple Flexible Wire Probes - KAPTON Insulated
Flexible Kapton-Insulated Probe for Faster Response Time than Most Insulated Wire Probes. Ideal for Checking Food Temperatures
  • Sold in packs of six.
  • Response Time: 0.35 seconds or greater
  • All insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue.
  • Custom designs or changes to this product are available upon request
Exposed-junction probes are not recommended for use with aqueous solutions.
Do not subject type J exposed-junction probes to moisture as they will rust.
19,100.00 - ₹21,600.00INR / Pkg of 6
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type Sheath Length (in)
Availability Pricing
COMPARE
500 T 60
₹21,600.00INR / Pkg of 6
COMPARE
600 K 60
₹19,100.00INR / Pkg of 6
COMPARE
600 J 60
₹21,300.00INR / Pkg of 6