Digi-Sense Thermocouple Flexible Wire Probes - Fiberglass-Insulated
Flexible Fiberglass Insulated Wire Used for High-Temperature Measurements
  • Response Time: 0.5 seconds or greater
  • All insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue.
  • Custom designs or changes to this product are available upon request
Fiberglass wire is not recommended for use in moist or wet environments.
Exposed-junction probes are not recommended for use with aqueous solutions.
Do not subject type J exposed-junction probes to moisture as they will rust
5,800.00 - ₹6,200.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type
Availability Pricing
COMPARE
900 J
₹5,900.00INR / Each
COMPARE
900 K
₹5,800.00INR / Each
COMPARE
500 T
₹6,200.00INR / Each