Digi-Sense Thermocouple Air/Gas Detachable Probes
Detachable Probes attach directly to your meter or probe handles
  • Probes Ideal for Measuring a Wide Range of Air Temperatures
  • Can be plugged directly into your meter.
  • Exposed Junction is protected by a rigid stainless steel shield to provide a fast response in an industrial environment
  • Response Time: 1.75 seconds or greater
  • Each probe comes with a color coded ANSI mini connector: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
7,300.00 - ₹7,500.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type
Availability Pricing
COMPARE
572 T
₹7,500.00INR / Each
COMPARE
572 K
₹7,300.00INR / Each