Conical-Bottom Open-Top Tanks with Stands
Tanks give you excellent chemical compatibility
Tanks are available in polypropylene or polyethylene; all stands are made from one-piece molded polyethylene. There are no metal parts—eliminate rust and corrosion problems.
308,000.00 - ₹617,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Max Temperature (° F) Volume (Gal)
Availability Pricing
Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PP, 55 gallon 0692908 Mfr # TC2247BP-FA0100-INSTALLED
COMPARE
Polypropylene 500 55
₹617,000.00INR / Each
Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PP, 250 gallon 0692918 Mfr # TC4257BP-FA0150-INSTALLED
COMPARE
Polypropylene 500 250
Call For Price
Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PE, 75 gallon 0692940 Mfr # TC3035AP-FA0150-INSTALLED
COMPARE
Polyethylene 75
₹308,000.00INR / Each
Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PE, 200 gallon 0692946 Mfr # TC3175AP-FA0150-INSTALLED
COMPARE
Polyethylene 200
₹370,000.00INR / Each