Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PE, 75 gallon
Representative image only.
Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PE, 75 gallon
Chem-Tainer | Mfr: TC3035AP-FA0150-INSTALLED| Item: 0692940
Tanks give you excellent chemical compatibility
₹308,000.00INR / Each
₹308,000.00INR / Each
Buy It Together
Current Item
Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PE, 75 gallon
Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PE, 75 gallon
0692940
₹308,000.00INR / Each
+
Chem-Tainer Low-Profile Cylindrical Tank, PE, 55 gallon
0692551
₹82,600.00INR / Each
=
Total Price 390600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Tank with a pre-drilled drain port and stand.
More About this Item
Tanks are available in polypropylene or polyethylene; all stands are made from one-piece molded polyethylene. There are no metal parts—eliminate rust and corrosion problems.
Specifications
  • MaterialPolyethylene
  • Volume (Gal)75
  • Volume (Liters)284
  • Height (in)35
  • Height (cm)88.9
  • Outside Diameter (OD) (in)30
  • DescriptionConical-Bottom Open-Top Tank with Stand, PE, 75 gallon