Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PP, 55 gallon
Representative image only.
Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PP, 55 gallon
Chem-Tainer | Mfr: TC2247BP-FA0100-INSTALLED| Item: 0692908
Tanks give you excellent chemical compatibility
₹617,000.00INR / Each
₹617,000.00INR / Each
Buy It Together
Current Item
Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PP, 55 gallon
Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PP, 55 gallon
0692908
₹617,000.00INR / Each
+
Chem-Tainer Low-Profile Cylindrical Tank, PE, 55 gallon
0692551
₹82,600.00INR / Each
=
Total Price 699600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Tank with a pre-drilled drain port and stand.
More About this Item
Tanks are available in polypropylene or polyethylene; all stands are made from one-piece molded polyethylene. There are no metal parts—eliminate rust and corrosion problems.
Specifications
  • MaterialPolypropylene
  • Volume (Gal)55
  • Volume (Liters)208
  • Height (in)47
  • Height (cm)119.38
  • Outside Diameter (OD) (in)22
  • Max Temperature (° F)500
  • Max Temperature (° C)260
  • DescriptionConical-Bottom Open-Top Tank with Stand, PP, 55 gallon