Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PE, 200 gallon
Representative image only.
Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PE, 200 gallon
Chem-Tainer | Mfr: TC3175AP-FA0150-INSTALLED| Item: 0692946
Tanks give you excellent chemical compatibility
₹370,000.00INR / Each
₹370,000.00INR / Each
Buy It Together
Current Item
Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PE, 200 gallon
Chem-Tainer Conical-Bottom Open-Top Tank with Stand, PE, 200 gallon
0692946
₹370,000.00INR / Each
+
Chem-Tainer Low-Profile Cylindrical Tank, PE, 55 gallon
0692551
₹82,600.00INR / Each
=
Total Price 452600.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Tank with a pre-drilled drain port and stand.
More About this Item
Tanks are available in polypropylene or polyethylene; all stands are made from one-piece molded polyethylene. There are no metal parts—eliminate rust and corrosion problems.
Specifications
  • MaterialPolyethylene
  • Volume (Gal)200
  • Volume (Liters)757
  • Height (in)75
  • Height (cm)190.5
  • Outside Diameter (OD) (in)31
  • DescriptionConical-Bottom Open-Top Tank with Stand, PE, 200 gallon