Cole-Parmer® Upright Freezer Racks for 0.2 mL Tube 96-Well PCR Boxes

Get organized for easier retrieval of samples
  • Locking rods keeps boxes securely in place
  • Easily access boxes from either side of rack
  • Sturdy corrosion-resistant stainless steel construction for long-term use
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors.
27,300.00 - ₹41,700.00INR / Each

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in) Length (in)
Availability Pricing
COMPARE
4 x 6 Array 8 15/16 14 11/16
₹30,900.00INR / Each
COMPARE
4 x 8 Array 11 7/8 14 11/16
₹39,700.00INR / Each
COMPARE
5 x 4 Array 6 18 3/8
₹27,300.00INR / Each
COMPARE
6 x 6 Array 8 15/16 22
₹41,700.00INR / Each
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat