Cole-Parmer® Upright Freezer Combination Drawer Racks for 2" Boxes and Centrifuge Tubes
Conveniently store both boxes and centrifuge tubes in a single rack
  • Use with standard 2" cardboard, plastic, or metal boxes and centrifuge tubes
  • Built-in labelling card holders help identify drawer content
  • Sturdy corrosion-resistant stainless steel construction
These specialized drawer racks lets you store 15 or 50-mL centrifuge tubes and storage bottles without a box. Drawer racks provide access to samples without having to remove the entire rack from the freezer.
38,300.00 - ₹56,500.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in) Length (in)
Availability Pricing
COMPARE
2 Drawers for 6 standard 2" boxes & 1 Drawer for Storage Bottles (drawer size: 15" x 4 1/2") 9 11/16 16 1/2
₹38,300.00INR / Each
COMPARE
2 Drawers for 8 standard 2" boxes & 1 Drawer for Storage Bottles (drawer size: 20 5/8" x 4 1/2") 9 11/16 22
₹42,100.00INR / Each
COMPARE
2 Drawers for 10 standard 2" boxes & 1 Drawer for Storage Bottles (drawer size: 25 1/2" x 4 1/2") 9 11/16 26 3/4
₹56,400.00INR / Each
COMPARE
2 Drawers for 6 standard 2" boxes & 1 Drawer for (60) 15-mL Centrifuge Tubes 9 11/16 16 1/2
₹39,700.00INR / Each
COMPARE
2 Drawers for 8 standard 2" boxes & 1 Drawer for (80) 15-mL Centrifuge Tubes 9 11/16 22
₹47,400.00INR / Each
COMPARE
2 Drawers for 6 standard 2" boxes & 1 Drawer for (30) 50-mL Centrifuge Tubes 9 11/16 16 1/2
₹39,200.00INR / Each
COMPARE
2 Drawers for 8 standard 2" boxes & 1 Drawer for (39) 50-mL Centrifuge Tubes 9 11/16 22
₹47,400.00INR / Each
COMPARE
2 Drawers for 10 standard 2" boxes & 1 Drawer for (51) 50-mL Centrifuge Tubes 9 11/16 26 3/4
₹56,500.00INR / Each
Showing 1- 8 of 8