Cole-Parmer® Thermocouple Compact Air/Gas Probes
Economical Air/Gas probes are ideal for quick readings
  • Exposed Junction is protected by a wire coil shield to minimize error from radiated heat
  • A compact light weight design allows for ease of use when space is limited but rugged enough for industrial environments.
  • Response Time: 1.75 seconds or greater
  • Each probe comes with a color coded ANSI coiled cable: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
8,200.00 - ₹8,600.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Probe Type
Availability Pricing
COMPARE
650 32 J
₹8,200.00INR / Each
COMPARE
650 -250 K
₹8,600.00INR / Each
COMPARE
572 -328 T
₹8,600.00INR / Each