Cole-Parmer® TELOS® Dry Loading Columns with Luer Lock Connection
Use dry loading columns with bulk sorbents to quickly and easily apply to flash chromatography systems
  • Columns are prefitted with a bottom frit
Attach dry loading columns directly to the main flash columns to suit majority of reaction scales. Columns feature luer lock inlet and luer slip outlet or luer lock inlet and outlet depending on model. Models with luer slip outlets can be used with ISCO CombiFlash® and Grace Reveleris® systems. Other models are best suited for SP and Idolera™ systems from Biotage®. They come pre-fitted with a bottom frit.
7,000.00 - ₹20,200.00INR / Pkg of 20
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Bed Mass (g) Fritted Disk
Availability Pricing
COMPARE
4 20 µm top frit, 10 µm bottom frit
₹12,500.00INR / Pkg of 20
COMPARE
12 20 µm top frit, 10 µm bottom frit
₹13,500.00INR / Pkg of 20
COMPARE
12 20 µm top frit, 10 µm bottom frit
₹17,600.00INR / Pkg of 20
COMPARE
20 20 µm top frit, 10 µm bottom frit
₹17,700.00INR / Pkg of 20
COMPARE
20 20 µm top frit, 10 µm bottom frit
₹20,200.00INR / Pkg of 20
COMPARE
40 20 µm top frit, 10 µm bottom frit
₹13,200.00INR / Pkg of 10
COMPARE
40 20 µm top frit, 10 µm bottom frit
₹11,100.00INR / Pkg of 10
COMPARE
45 20 µm top frit, 10 µm bottom frit
₹12,500.00INR / Pkg of 10
COMPARE
80 20 µm top frit, 10 µm bottom frit
₹7,000.00INR / Pkg of 5
COMPARE
120 20 µm top frit, 10 µm bottom frit
₹14,000.00INR / Pkg of 5
COMPARE
330 20 µm top frit, 10 µm bottom frit
₹9,700.00INR / Pkg of 1
COMPARE
4 10 µm bottom frit, no top frit
₹13,000.00INR / Pkg of 20
COMPARE
12 10 µm bottom frit, no top frit
₹14,900.00INR / Pkg of 20
COMPARE
20 10 µm bottom frit, no top frit
₹16,900.00INR / Pkg of 20
COMPARE
40 10 µm bottom frit, no top frit
₹11,100.00INR / Pkg of 10
COMPARE
80 10 µm bottom frit, no top frit
₹7,800.00INR / Pkg of 5
Showing 1- 16 of 16