Cole-Parmer TELOS® Dry Loading Cartridge Flash Column, 10 µm frit, 80g, slip tip; 5/Pk
Representative image only.
Cole-Parmer TELOS® Dry Loading Cartridge Flash Column, 10 µm frit, 80g, slip tip; 5/Pk
Cole-Parmer | Item: 0647571
Use dry loading columns with bulk sorbents to quickly and easily apply to flash chromatography systems
₹7,800.00INR / Pkg of 5
₹7,800.00INR / Pkg of 5
Product Description
Key Features
  • Columns are prefitted with a bottom frit
  • No top frit
More About this Item
Attach dry loading columns directly to the main flash columns to suit majority of reaction scales. Columns feature luer lock inlet and luer slip outlet or luer lock inlet and outlet depending on model. Models with luer slip outlets can be used with ISCO CombiFlash® and Grace Reveleris® systems. Other models are best suited for SP and Idolera™ systems from Biotage®. They come pre-fitted with a bottom frit.
Specifications
  • Bed Mass (g)80
  • Fritted Disk10 µm bottom frit, no top frit
  • Compatable WithISCO CombiFlash® and Grace Reveleris®
  • ConnectionsLuer slip tip inlet / outlet
  • DescriptionDry Loading Cartridge, 10 µm Bottom Frit, Luer Lock Inlet/Luer Slip Outlet, 80 g; 5/Pk