Cole-Parmer TELOS® Dry Loading Cartridge Screw-on Flash Column, 20 µm top frit, 10 µm bottom frit 40g, Luer Lock; 10/Pk
Representative image only.
Cole-Parmer TELOS® Dry Loading Cartridge Screw-on Flash Column, 20 µm top frit, 10 µm bottom frit 40g, Luer Lock; 10/Pk
Cole-Parmer | Item: 0647559
Use dry loading columns with bulk sorbents to quickly and easily apply to flash chromatography systems
₹13,200.00INR / Pkg of 10
₹13,200.00INR / Pkg of 10
Buy It Together
Current Item
Cole-Parmer TELOS® Dry Loading Cartridge Screw-on Flash Column, 20 µm top frit, 10 µm bottom frit 40g, Luer Lock; 10/Pk
Cole-Parmer TELOS® Dry Loading Cartridge Screw-on Flash Column, 20 µm top frit, 10 µm bottom frit 40g, Luer Lock; 10/Pk
0647559
₹13,200.00INR / Pkg of 10
+ =
Total Price 33600.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Columns are prefitted with a bottom frit
More About this Item
Attach dry loading columns directly to the main flash columns to suit majority of reaction scales. Columns feature luer lock inlet and luer slip outlet or luer lock inlet and outlet depending on model. Models with luer slip outlets can be used with ISCO CombiFlash® and Grace Reveleris® systems. Other models are best suited for SP and Idolera™ systems from Biotage®. They come pre-fitted with a bottom frit.
Specifications
  • Bed Mass (g)40
  • Fritted Disk20 µm top frit, 10 µm bottom frit
  • Compatable WithSP and Idolera™ systems from Biotage®
  • DescriptionDry Loading Cartridge, 20 µm Top Frit, 10 µm Bottom Frit, Luer Lock Inlet/Outlet, 40 g; 10/Pk