Cole-Parmer TELOS® Dry Loading Cartridge Screw-on Flash Column, 20 µm top frit, 10 µm bottom frit, 20g, Luer Lock; 20/Pk
Representative image only.
Cole-Parmer TELOS® Dry Loading Cartridge Screw-on Flash Column, 20 µm top frit, 10 µm bottom frit, 20g, Luer Lock; 20/Pk
Cole-Parmer | Item: 0647556
Use dry loading columns with bulk sorbents to quickly and easily apply to flash chromatography systems
₹17,700.00INR / Pkg of 20
₹17,700.00INR / Pkg of 20
Buy It Together
Current Item
Cole-Parmer TELOS® Dry Loading Cartridge Screw-on Flash Column, 20 µm top frit, 10 µm bottom frit, 20g, Luer Lock; 20/Pk
Cole-Parmer TELOS® Dry Loading Cartridge Screw-on Flash Column, 20 µm top frit, 10 µm bottom frit, 20g, Luer Lock; 20/Pk
0647556
₹17,700.00INR / Pkg of 20
+ =
Total Price 41100.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Columns are prefitted with a bottom frit
More About this Item
Attach dry loading columns directly to the main flash columns to suit majority of reaction scales. Columns feature luer lock inlet and luer slip outlet or luer lock inlet and outlet depending on model. Models with luer slip outlets can be used with ISCO CombiFlash® and Grace Reveleris® systems. Other models are best suited for SP and Idolera™ systems from Biotage®. They come pre-fitted with a bottom frit.
Specifications
  • Bed Mass (g)20
  • Fritted Disk20 µm top frit, 10 µm bottom frit
  • Compatable WithSP and Idolera™ systems from Biotage®
  • DescriptionDry Loading Cartridge, 20 µm Top Frit, 10 µm Bottom Frit, Luer Lock Inlet/Outlet, 20 g; 20/Pk