Cole-Parmer® Pestles, Porcelain
Oversize pestle provides comfort while grinding
  • Thermal shock, chemical and heat resistant
  • Autoclavable
The pestles are suitable for basic laboratory purposes, sample preparation, and emulsifications. An oversized pestle offers hand comfort while grinding samples. Pestles are glazed all over except for grinding surfaces.
2,700.00 - ₹62,500.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Pestles Length (mm) Description
Availability Pricing
COMPARE
114 Pestle, Porcelain, 114 mm Length
₹2,700.00INR / Each
COMPARE
157 Pestle, Porcelain, 157 mm Length
₹2,900.00INR / Each
COMPARE
180 Pestle, Porcelain, 180 mm Length
₹3,400.00INR / Each
COMPARE
194 Pestle, Porcelain, 194 mm Length
₹5,000.00INR / Each
COMPARE
222 Pestle, Porcelain, 222 mm Length
₹7,900.00INR / Each
COMPARE
262 Pestle, Porcelain, 262 mm Length
₹12,100.00INR / Each
COMPARE
130 Pestle, Porcelain; 130 mm Length; 18/PK
₹47,700.00INR / Pkg of 18
COMPARE
157 Pestle, Porcelain; 157 mm Length; 12/PK
₹62,500.00INR / Pkg of 12
COMPARE
180 Pestle, Porcelain; 180 mm Length; 8/PK
₹26,500.00INR / Pkg of 8
COMPARE
222 Pestle, Porcelain; 222 mm Length; 4/PK
₹35,300.00INR / Pkg of 4
COMPARE
262 Pestle, Porcelain; 262 mm Length; 2/PK
₹24,200.00INR / Pkg of 2
Showing 1- 11 of 11