Cole-Parmer® In-line or Submersible, Solution Grounded pH/ORP Electrodes
Industrial pH/ORP Electrodes for Chemical Property Testing
  • Get longer life with PTFE double junctions
  • Ideal for harsh conditions
These electrodes offer solution ground, PTFE references with double junctions and polymerized gel. All models include a 10-ft cable with BNC pH connector and Ryton® housing with 3/4'' NPT(M) threads.
46,300.00 - ₹93,500.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Solution Grounded pH Probe, DJ/PPS/No ATC; BNC
₹46,300.00INR / Each
COMPARE
Solution Grounded pH Probe, DJ/PPS/100Ohm RTD; Plug/BNC
₹93,500.00INR / Each
COMPARE
Solution Grounded pH Probe, DJ/PPS/100Ohm RTD; Tinned/BNC
₹59,700.00INR / Each
COMPARE
Solution Grounded High-Na pH Probe, DJ/PPS/No ATC; BNC
₹46,300.00INR / Each
COMPARE
Solution Grounded High-Na pH Probe, DJ/PPS/100Ohm RTD; Plug/BNC
₹58,200.00INR / Each
COMPARE
Solution Grounded High-Na pH Probe, DJ/PPS/100Ohm RTD; Tinned/BNC
₹56,400.00INR / Each
COMPARE
Solution Grounded High-Na pH Probe, DJ/PPS/1kOhm RTD; Spade/BNC
₹54,800.00INR / Each
COMPARE
Solution Grounded, Platinum-Band ORP Electrode; No ATC Element, BNC
₹51,800.00INR / Each
COMPARE
Solution Grounded, Gold-Band ORP Electrode; No ATC Element, BNC
₹53,900.00INR / Each
Showing 1- 9 of 9