Cole-Parmer Solution Grounded pH Probe, DJ/PPS/No ATC; BNC
Representative image only.
Cole-Parmer Solution Grounded pH Probe, DJ/PPS/No ATC; BNC
Cole-Parmer | Item: 2701800
Industrial pH/ORP Electrodes for Chemical Property Testing
₹46,300.00INR / Each
₹46,300.00INR / Each
Buy It Together
Current Item
Cole-Parmer Solution Grounded pH Probe, DJ/PPS/No ATC; BNC
Cole-Parmer Solution Grounded pH Probe, DJ/PPS/No ATC; BNC
2701800
₹46,300.00INR / Each
+
Cole-Parmer Insertion/Submersion ORP Probe; DJ/CPVC/; /BNC
2730119
₹44,400.00INR / Each
+ =
Total Price 149000.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Get longer life with PTFE double junctions
  • Ideal for harsh conditions
  • 1 year warranty
More About this Item
These electrodes offer solution ground, PTFE references with double junctions and polymerized gel. All models include a 10-ft cable with BNC pH connector and Ryton® housing with 3/4'' NPT(M) threads.
General-Purpose Electrodes have low-resistant glass with a very fast and stable response. Designed for minimal salt applications. Use for measurements from 0 to 13 pH. Temperature range is 32 to 212°F (0 to 100°C).
Keep a Spare1 Year Warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialRyton® PPS
  • Min pH (pH)13
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • ATC ElementNone
  • ConnectorBNC
  • DescriptionSolution Grounded pH Probe, DJ/PPS/No ATC; BNC