Cole-Parmer Insertion/Submersion ORP Probe; DJ/CPVC/; /BNC
Representative image only.
Cole-Parmer Insertion/Submersion ORP Probe; DJ/CPVC/; /BNC
Cole-Parmer | Item: 2730119
Reduce bulb damage with probe guard
₹44,400.00INR / Each
₹44,400.00INR / Each
Buy It Together
Current Item
Cole-Parmer Insertion/Submersion ORP Probe; DJ/CPVC/; /BNC
Cole-Parmer Insertion/Submersion ORP Probe; DJ/CPVC/; /BNC
2730119
₹44,400.00INR / Each
+ + =
Total Price 130400.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Versatile design—threads on both ends for submersion or in-line use Double-junction electrodes available; resist contamination from heavy metals, organics, or AgCI reactive compounds
  • 6 month warranty
More About this Item
In-line electrodes with probe guard operate from 23 to 176°>F (-5 to 80°>C) and have a PVC housing with a 3/4'' NPT(M) fitting on both ends. Use as a submersion sensor. Epoxy body withstands pressures to 100 psi. Measure pH from 0 to 14. ORP electrode impedance is 100 MΩ. All electrodes measure 6-3/8''L x 1'' OD.
6 Month Warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialPVC
  • Min pH (pH)14
  • Max Temperature (° C)80
  • ConnectorBNC
  • DescriptionInsertion/Submersion ORP Probe; DJ/CPVC/; /BNC
  • Warranty6 month warranty