Cole-Parmer In-Line pH Electrode with Tuff-Tip, No ATC Element
Representative image only.
Cole-Parmer In-Line pH Electrode with Tuff-Tip, No ATC Element
Cole-Parmer | Item: 0599501
Semi-domed, tuff-tip limits glass exposure and protects electrode from damage
₹29,600.00INR / Each
₹29,600.00INR / Each
Buy It Together
Current Item
Cole-Parmer In-Line pH Electrode with Tuff-Tip, No ATC Element
Cole-Parmer In-Line pH Electrode with Tuff-Tip, No ATC Element
0599501
₹29,600.00INR / Each
+ + + +
Cole-Parmer Insertion/Submersion ORP Probe; DJ/CPVC/; /BNC
2730119
₹44,400.00INR / Each
=
Total Price 400800.00 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Order single or double junction disposable electrodes for your in-line process control needs
  • Available with BNC or tinned ends
  • 1 year warranty
More About this Item
Electrodes feature PVDF housing with 1/2'' NPT(M) threads and withstands temperatures from 23 to 176°F (--5 to 80°C) and pressures to 100 psi. Electrodes include 10-ft cable with BNC pH connector and tinned ends for ATC element. Insertion depth 1''.
Keep a Spare1 Year Warranty
Specifications
  • ApplicationsIn-line
  • Electrode TypeSingle junction
  • Electrode Body MaterialPVDF
  • Min Temperature (° C)-5
  • Max Temperature (° C)80
  • GuardTuff-Tip
  • Thread Size NPT(M) (in)1/2
  • ConnectorBNC
  • Cable Length (m)3
  • DescriptionIn-Line pH Electrode with Tuff-Tip, No ATC Element