Cole-Parmer® Tuff-Tip Bulb, In-line pH and ORP Electrodes
Semi-domed, tuff-tip limits glass exposure and protects electrode from damage
  • Order single or double junction disposable electrodes for your in-line process control needs
  • Available with BNC or tinned ends
Electrodes feature PVDF housing with 1/2'' NPT(M) threads and withstands temperatures from 23 to 176°F (--5 to 80°C) and pressures to 100 psi. Electrodes include 10-ft cable with BNC pH connector and tinned ends for ATC element. Insertion depth 1''.
29,600.00 - ₹46,200.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
In-Line pH Electrode with Tuff-Tip, No ATC Element
₹29,600.00INR / Each
COMPARE
Tuff-Tip In-Line pH Probe, DJ/PVDF/; /BNC
₹34,300.00INR / Each
COMPARE
Tuff-Tip In-Line pH Probe, DJ/PVDF/1kOhm RTD; Spade/BNC
₹40,800.00INR / Each
COMPARE
Tuff-Tip In-Line ORP Probe, DJ/PVDF/10'/BNC
₹46,200.00INR / Each