Cole-Parmer Tuff-Tip In-Line pH Probe, DJ/PVDF/1kOhm RTD; Spade/BNC
Representative image only.
Cole-Parmer Tuff-Tip In-Line pH Probe, DJ/PVDF/1kOhm RTD; Spade/BNC
Cole-Parmer | Item: 0599514
Semi-domed, tuff-tip limits glass exposure and protects electrode from damage
₹40,800.00INR / Each
₹40,800.00INR / Each
Buy It Together
Current Item
Cole-Parmer Tuff-Tip In-Line pH Probe, DJ/PVDF/1kOhm RTD; Spade/BNC
Cole-Parmer Tuff-Tip In-Line pH Probe, DJ/PVDF/1kOhm RTD; Spade/BNC
0599514
₹40,800.00INR / Each
+ +
Cole-Parmer Insertion/Submersion ORP Probe; DJ/CPVC/; /BNC
2730119
₹44,400.00INR / Each
+ =
Total Price 396100.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Order single or double junction disposable electrodes for your in-line process control needs
  • Available with BNC or tinned ends
  • 1 year warranty
More About this Item
Electrodes feature PVDF housing with 1/2'' NPT(M) threads and withstands temperatures from 23 to 176°F (--5 to 80°C) and pressures to 100 psi. Electrodes include 10-ft cable with BNC pH connector and tinned ends for ATC element. Insertion depth 1''.
Keep a Spare1 Year Warranty
Specifications
  • ApplicationsIn-line
  • Electrode TypeDouble junction
  • Electrode Body MaterialPVDF
  • Min Temperature (° C)-5
  • Max Temperature (° C)80
  • Temperature CompensationYes
  • GuardTuff-Tip
  • Thread Size NPT(M) (in)1/2
  • ATC ConnectorSpade lugs
  • ATC Element1 kΩ Pt RTD
  • ConnectorBNC
  • Cable Length (m)3
  • DescriptionTuff-Tip In-Line pH Probe, DJ/PVDF/1kOhm RTD; Spade/BNC