Cole-Parmer® Self-Cleaning, Flat Surface ORP Electrodes
Flat electrode surface prevents buildup of solids
  • Flat surface prevents buildups, while fluid flow cleans surface for longer life Use with most meters and controllers Double junction for difficult applications Choose either CPVC housing or chemical-resistant Ryton® PPS housing
Use these unique double-junction electrodes in your most difficult ORP applications. Flat surface does not collect suspended solids--fluid flow across the electrode provides a cleaning action that extends electrode life and improves performance. Annular junction reduces reference cell fouling. Choose either a CPVC housing (max temp 80°>C) or chemical resistant Ryton® housing (max temp 100°>C). Both types have 10-ft cables and can be used at pressures up to 100 psi. BNC connectors match a variety of meters and controllers. Install electrodes in tee fittings--3/4'' NPT(M) thread and 2'' insertion depth.
37,900.00 - ₹44,700.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Self-Cleaning Flat Surface ORP Probe, DJ/CPVC/Pt/10'/BNC
₹37,900.00INR / Each
COMPARE
Self-Cleaning Flat Surface ORP Probe, DJ/Ryton/Pt/10'/BNC
₹40,800.00INR / Each
COMPARE
Self-Cleaning Flat Surface ORP Probe, DJ/Ryton/Au/10'/BNC
₹44,700.00INR / Each