Cole-Parmer Solution Grounded, Gold-Band ORP Electrode; No ATC Element, BNC
Representative image only.
Cole-Parmer Solution Grounded, Gold-Band ORP Electrode; No ATC Element, BNC
Cole-Parmer | Item: 2701850
Industrial pH/ORP Electrodes for Chemical Property Testing
₹53,900.00INR / Each
₹53,900.00INR / Each
Buy It Together
Current Item
Cole-Parmer Solution Grounded, Gold-Band ORP Electrode; No ATC Element, BNC
Cole-Parmer Solution Grounded, Gold-Band ORP Electrode; No ATC Element, BNC
2701850
₹53,900.00INR / Each
+ +
Cole-Parmer Insertion/Submersion ORP Probe; DJ/CPVC/; /BNC
2730119
₹44,400.00INR / Each
=
Total Price 156600.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Get longer life with PTFE double junctions
  • Ideal for harsh conditions
  • 1 year warranty
More About this Item
These electrodes offer solution ground, PTFE references with double junctions and polymerized gel. All models include a 10-ft cable with BNC pH connector and Ryton® housing with 3/4'' NPT(M) threads.
Platinum/Gold-Ring ORP Electrodes have a metal-band sensor for oxidation-reduction measurements. Temperature range is 32 to 266°F (0 to 130°C).
Keep a Spare1 Year Warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialRyton® PPS
  • Reference JunctionPTFE
  • Reference ElectrolytePolymerized gel
  • Min Temperature (° C)0
  • Max Temperature (° C)130
  • ATC ElementNone
  • ConnectorBNC
  • DescriptionSolution Grounded, Gold-Band ORP Electrode; No ATC Element, BNC