Cole-Parmer Solution Grounded High-Na pH Probe, DJ/PPS/100Ohm RTD; Plug/BNC
Representative image only.
Cole-Parmer Solution Grounded High-Na pH Probe, DJ/PPS/100Ohm RTD; Plug/BNC
Cole-Parmer | Item: 2701822
Industrial pH/ORP Electrodes for Chemical Property Testing
₹58,200.00INR / Each
₹58,200.00INR / Each
Buy It Together
Current Item
Cole-Parmer Solution Grounded High-Na pH Probe, DJ/PPS/100Ohm RTD; Plug/BNC
Cole-Parmer Solution Grounded High-Na pH Probe, DJ/PPS/100Ohm RTD; Plug/BNC
2701822
₹58,200.00INR / Each
+
Cole-Parmer Insertion/Submersion ORP Probe; DJ/CPVC/; /BNC
2730119
₹44,400.00INR / Each
+ =
Total Price 160900.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Get longer life with PTFE double junctions
  • Ideal for harsh conditions
  • 1 year warranty
More About this Item
These electrodes offer solution ground, PTFE references with double junctions and polymerized gel. All models include a 10-ft cable with BNC pH connector and Ryton® housing with 3/4'' NPT(M) threads.
High-Salt Application Electrodes are designed for continuous long-term use at high temperatures in strong alkaline solutions above pH 11. Use for measurements from 0 to 14 pH. Negligible salt ion error may be expected above pH 13. Response time will increase as solution temperature rises due to high glass impedance. Temperature range is 32 to 275°F (0 to 135°C).
Keep a Spare1 Year Warranty
Specifications
  • Electrode TypeDouble junction
  • Electrode Body MaterialRyton® PPS
  • Min pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)135
  • Temperature CompensationYes
  • ATC ConnectorSpade lugs
  • ATC Element100 Ω Pt RTD
  • ConnectorBNC
  • DescriptionSolution Grounded High-Na pH Probe, DJ/PPS/100Ohm RTD; Plug/BNC