Cole-Parmer® Heavy-Duty Plastic Bottles, PP
Withstands repeated full vacuum applications
  • Hold full vacuum for up to 24 hours
  • Polypropylene closure with thermoplastic
  • Autoclavable
These bottles can withstand repeated application of full vacuum for 24 hours at 20°C (68°F). The polypropylene (PP) closure features a thermoplastic elastomer (TPE) gasket—use as a waste aspirator when fitted with a filling/venting cap or barbed bulkhead fitting kit (sold separately).
13,400.00 - ₹39,400.00INR / Pkg of 2
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
COMPARE
4 83B Polypropylene(PP)
₹13,400.00INR / Each
COMPARE
0.25 53B Polypropylene(PP)
₹22,100.00INR / Pkg of 6
COMPARE
0.5 53B Polypropylene(PP)
₹29,200.00INR / Pkg of 6
COMPARE
1 53B Polypropylene(PP)
₹39,400.00INR / Pkg of 6
COMPARE
2 53B Polypropylene(PP)
₹16,200.00INR / Pkg of 2
COMPARE
5 83B Polypropylene(PP)
₹17,500.00INR / Each