Cole-Parmer® Economy Amber Wide-Mouth Plastic Bottle, HDPE
Keep light-sensitive samples safe
  • Reduce UV light transmission
  • Meets USP Class VI requirements
  • Will not leak with PP screw caps
Protect light-sensitive materials during shipping and storage. Amber bottles reduce UV light transmission and meet USP requirements for storage of light-sensitive materials. Suitable for packing, storage, and shipping liquids. All have leakproof polypropylene screw caps.
3,900.00 - ₹37,000.00INR / Pkg of 12
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
30 28 mm High Density Polyethyene (HDPE)
₹3,900.00INR / Pkg of 12
COMPARE
30 28 mm High Density Polyethyene (HDPE)
₹21,200.00INR / Pkg of 72
COMPARE
60 28 mm High Density Polyethyene (HDPE)
₹3,900.00INR / Pkg of 12
COMPARE
60 28 mm High Density Polyethyene (HDPE)
₹21,400.00INR / Pkg of 72
COMPARE
125 38 mm High Density Polyethyene (HDPE)
₹4,400.00INR / Pkg of 12
COMPARE
125 38 mm High Density Polyethyene (HDPE)
₹24,500.00INR / Pkg of 72
COMPARE
250 43 mm High Density Polyethyene (HDPE)
₹6,600.00INR / Pkg of 12
COMPARE
250 43 mm High Density Polyethyene (HDPE)
₹37,000.00INR / Pkg of 72
COMPARE
500 53 mm High Density Polyethyene (HDPE)
₹8,600.00INR / Pkg of 12
COMPARE
500 53 mm High Density Polyethyene (HDPE)
₹32,300.00INR / Pkg of 48
COMPARE
1000 63 mm High Density Polyethyene (HDPE)
₹7,800.00INR / Pkg of 6
COMPARE
1000 63 mm High Density Polyethyene (HDPE)
₹29,100.00INR / Pkg of 24
Showing 1- 12 of 12